EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
Nanoengineered porous chitosan/CaTiO
PMID 28672204
Abstract
To further improve the property of promoting peripheral nerve regeneration of chitosan materials, CaTiO