EMAIL THIS PAGE TO A FRIEND
Nutrients
N-3 Polyunsaturated Fatty Acids Decrease the Protein Expression of Soluble Epoxide Hydrolase via Oxidative Stress-Induced P38 Kinase in Rat Endothelial Cells.
PMID 28672788