EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Small interfering RNA targeting of the survivin gene inhibits human tumor cell growth
PMID 28672890
Abstract
The present study aimed to evaluate the impact of small interfering RNA (siRNA) targeting of the survivin gene in human tumor cells and the effect of decreased survivin expression on the proliferation and apoptosis of tumor cells. Human tumor cell lines (MSA-MB-231, SGC-7901, HeLa, A549, SK-OV-3 and Raji, PC-3) were cultured