EMAIL THIS PAGE TO A FRIEND
PMID 28672947
Abstract
The constituents of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4301218
Anti-HSPG2 antibody produced in rabbit, affinity isolated antibody