EMAIL THIS PAGE TO A FRIEND
Frontiers in chemistry
Allosteric Control of Substrate Specificity of the
PMID 28674689
Abstract
The substrate specificity of enzymes is crucial to control the fate of metabolites to different pathways. However, there is growing evidence that many enzymes can catalyze alternative reactions. This promiscuous behavior has important implications in protein evolution and the acquisition of new functions. The question is how the undesirable outcomes of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0627
Adenosine-5′-diphosphoglucose disodium salt, ≥93%
C16H23N5Na2O15P2