EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Pd-Catalyzed Carbonylative Carboperfluoroalkylation of Alkynes. Through-Space 13C-19F Coupling as a Probe for Configuration Assignment of Fluoroalkyl-Substituted Olefins.
PMID 28677978
Abstract
A four-component Pd-catalyzed protocol for direct synthesis of perfluoroalkyl-substituted enones is reported. Under mild conditions and low catalyst loading, alkynes, iodoperfluoroalkanes, (hetero)arylboronic acids, and carbon monoxide are assembled into highly elaborate products with good yields and excellent regio- and stereoselectivities. The configuration of the products was confirmed by the observation of through-space 13C-19F couplings, accessible through the analysis of routine 13C NMR spectra.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
905607
(R)-Tol-BINAP Pd G3, ≥95%
C61H53NO3P2PdS