EMAIL THIS PAGE TO A FRIEND
Journal of medicinal food
Aronia Berry Extract Ameliorates the Severity of Dextran Sodium Sulfate-Induced Ulcerative Colitis in Mice.
PMID 28677983
Abstract
Inflammatory bowel disease, including Crohn's disease and ulcerative colitis (UC), is a group of inflammatory conditions of the colon and small intestine. UC is a chronic inflammatory disorder of the colon and rectum that includes intervals of acute exacerbation. Although recent studies have suggested that proinflammatory cytokines might have initiated the inflammatory responses in UC, its etiology remains unclear. Aronia berries are rich in dietary polyphenols such as phenolic acids, anthocyanins, flavonoids, and proanthocyanidins with various health benefits, including antioxidant, anti-inflammatory, and antiaging activities. The objective of this study was to determine whether Aronia berry can be an effective intervention for the treatment of UC. BALB/c mice were administered 5% dextran sulfate sodium (DSS) to induce UC. They were then given Aronia berry extracts at concentrations of 10 or 100 mg/kg. During the induction of UC, the expression levels of nuclear factor-kappa B were increased in colonic epithelial cells and immune cells, leading to increased proinflammatory cytokine levels. Aronia berry extract significantly improved the clinical signs of DSS-induced UC, including body weight loss, colon length shortening, and disease activity index increase, with histological markers of colon injury. Furthermore, oral administration of Aronia berry extract inhibited prostaglandin E