EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Raman spectral titration method: an informative technique for studying the complexation of uranyl with uranyl(vi)-DPA/oxalate systems as examples.
PMID 28678293
Abstract
The Raman band at about 870 cm