EMAIL THIS PAGE TO A FRIEND
Chemosphere
Non-target evaluation of contaminants in honey bees and pollen samples by gas chromatography time-of-flight mass spectrometry.
PMID 28679151
Abstract
This work presents a non-targeted screening approach for the detection and quantitation of contaminants in bees and pollen, collected from the same hive, by GC-EI-ToF-MS. It consists of a spectral library datasets search using a compound database followed by a manual investigation and analytical standard confirmation together with semi-quantitation purposes. Over 20% of the compounds found automatically by the library search could not be confirmed manually. This number of false positive detections was mainly a consequence of an inadequate ion ratio criterion (±30%), not considered in the automatic searching procedure. Eight compounds were detected in bees and pollen. They include insecticides/acaricides (chlorpyrifos, coumaphos, fluvalinate-tau, chlorfenvinphos, pyridaben, and propyl cresol) at a concentration range of 1-1207 μg kg