EMAIL THIS PAGE TO A FRIEND
Water research
Bromate inhibition by reduced graphene oxide in thermal/PMS process.
PMID 28679477
Abstract
Bromate (BrO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
228036
OXONE®, monopersulfate compound
HKO5S · 0.5HKO4S · 0.5K2O4S