EMAIL THIS PAGE TO A FRIEND
Journal of virology
AAV serotypes have distinctive interactions with domains of the cellular receptor AAVR.
PMID 28679762
Abstract
Adeno-associated virus (AAV) entry is determined by its interactions with specific surface glycans and proteinaceous receptor(s). Adeno-associated virus receptor (AAVR; also named KIAA0319L) is an essential cellular receptor required for the transduction of vectors derived from multiple AAV serotypes including the evolutionary distant serotypes, AAV2 and AAV5. Here, we further biochemically characterize the AAV-AAVR interaction and define the domains within the ectodomain of AAVR that facilitate this interaction. Using a virus overlay assay, it was previously shown that the major AAV2 binding protein in membrane preparations of human cells corresponds to a glycoprotein with a 150-kDa molecular mass. By establishing a purification procedure, performing further protein separation through two-dimensional electrophoresis and utilizing mass spectrometry, we now show that this glycoprotein is identical to AAVR. While we find that AAVR is N-linked glycosylated, this glycosylation is not a strict requirement for AAV2 binding or functional transduction. Using a combination of genetic complementation with deletion constructs and viral overlay assays with individual domains, we find that AAV2 functionally interacts predominantly with the second Ig-like PKD repeat domain (PKD2) present in the ectodomain of AAVR. By contrast, AAV5 interacts primarily through the first, most membrane distal, PKD domain (PKD1) of AAVR to promote transduction. Furthermore, other AAV serotypes including AAV1 and 8 require a combination of PKD1 and PKD2 for optimal transduction. These results suggest that despite their shared dependence on AAVR as a critical entry receptor, different AAV serotypes have evolved distinctive interactions with the same receptor.