EMAIL THIS PAGE TO A FRIEND
The Journal of experimental biology
The Ol
PMID 28679796
Abstract
Mapping brain function to brain structure is a fundamental task for neuroscience. For such an endeavour, the
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0463
BPD, ≥98% (HPLC)
C17H10O5