EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
IL-4 up-regulates cyclooxygenase-1 expression in macrophages.
PMID 28684424
Abstract
Macrophages use various cell-surface receptors to sense their environment and undergo polarized responses. The cytokines, interleukin (IL)-4 and IL-13, released from T-helper type 2 (Th2) cells, drive macrophage polarization toward an alternatively activated phenotype (M2). This phenotype is associated with the expression of potent pro-resolving mediators, such as the prostaglandin (PG) D