EMAIL THIS PAGE TO A FRIEND
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Neuroprotective effect of Indian propolis in β-amyloid induced memory deficit: Impact on behavioral and biochemical parameters in rats.
PMID 28686968
Abstract
The study aimed at the investigation of neuroprotective activity of macerated ethanolic extract of Indian propolis (MEEP) against β-Amyloid 25-35 (Aβ
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282200
Galangin, autophagy inducing flavonoid
C15H10O5