EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Propylsulfonic acid-anchored isocyanurate-based periodic mesoporous organosilica (PMO-ICS-Pr-SO
PMID 28687033
Abstract
A new propylsulfonic acid-anchored isocyanurate bridging periodic mesoporous organosilica (PMO-ICS-Pr-SO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282626
Trimethoxysilane, 95%
C3H10O3Si