EMAIL THIS PAGE TO A FRIEND
Mycotoxin research
Use of yeast (Pichia kudriavzevii) as a novel feed additive to ameliorate the effects of aflatoxin B
PMID 28687999
Abstract
The aim of this study was to evaluate the efficacy of autochthonous Pichia kudriavzevii as a novel bioadsorbent for aflatoxin B