EMAIL THIS PAGE TO A FRIEND
Oncology letters
Inhibition of paclitaxel resistance and apoptosis induction by cucurbitacin B in ovarian carcinoma cells.
PMID 28693146
Abstract
Ovarian cancer is the leading cause of mortality among all gynecological malignancies. Drug resistance is a cause of ovarian cancer recurrence and low rate of overall survival. There is a requirement for more effective treatment approaches. Cucurbitacin B (CuB) is an antineoplastic agent derived from traditional Chinese medicinal herbs. Its activity against paclitaxel-resistant human ovarian cancer cells has, however, not yet been established. The purpose of the present study was to investigate the effect and mechanism of CuB on human paclitaxel-resistant ovarian cancer A2780/Taxol cells. Cell viability was evaluated by a cell counting assay, while cell cycle arrest and apoptosis were assessed by microscopy and flow cytometry, and proteins associated with apoptotic pathways and drug resistance were evaluated by western blotting. The present results demonstrated that CuB exerts dose- and time-dependent cytotoxicity against the ovarian cancer A2780 cell line, with half-maximal inhibitory concentration (IC