EMAIL THIS PAGE TO A FRIEND
Oncology letters
Monocyte chemoattractant protein 1 expression and proliferation in primary central nervous system lymphoma.
PMID 28693163
Abstract
Whether the poor prognosis of primary central nervous system lymphoma (PCNSL) compared with systemic diffuse large B cell lymphoma (DLBCL) is attributable to the immune privilege of the intracerebral location or to intrinsic differences in the biological characteristics of two types of lymphoma remains unclear. Monocyte chemoattractant protein 1 (MCP-1) is essential to support tumor cell survival and growth, and the present study aimed to compare MCP-1 expression in PCNSL and peripheral DLBCL. The present study included 19 patients with PCNSL and 16 patients with DLBCL, all of whom had tissue diagnosis and lymphoma tissue samples available for analysis. Histology included immunohistochemistry using antibodies against a panel of lymphoma markers, antibodies specific to MCP-1, and antibodies specific to tumor-associated macrophages. MCP-1 expression was quantified using immunostaining scoring. RNA extraction and reverse transcription-quantitative polymerase chain reaction were used to determine