EMAIL THIS PAGE TO A FRIEND
Oncology letters
Transforming growth factor-β1 contributes to oxaliplatin resistance in colorectal cancer via epithelial to mesenchymal transition.
PMID 28693217
Abstract
Transforming growth factor-β1 (TGF-β1), secreted by main components of tumor microenvironment, is considered to be closely associated with cancer development and chemoresistance. The present study aimed to analyze the effects and mechanisms underlying TGF-β1-induced chemoresistance to oxaliplatin (LOH) in human colorectal cancer (CRC) cell lines. The cytotoxic effects of LOH subsequent to TGF-β1 treatment were assessed in three CRC cell lines by MTT assay. In addition, epithelial to mesenchymal transition (EMT), DNA damage and apoptosis assays were performed to evaluate the mechanisms of drug resistance