EMAIL THIS PAGE TO A FRIEND
Frontiers in pharmacology
The Effect of Pungent and Tingling Compounds from
PMID 28694780
Abstract
Black peppercorns (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C191
Capsazepine, ≥98% (HPLC), solid
C19H21ClN2O2S