EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Analytical methodologies for broad metabolite coverage of exhaled breath condensate.
PMID 28697414
Abstract
Breath analysis has been gaining popularity as a non-invasive technique that is amenable to a broad range of medical uses. One of the persistent problems hampering the wide application of the breath analysis method is measurement variability of metabolite abundances stemming from differences in both sampling and analysis methodologies used in various studies. Mass spectrometry has been a method of choice for comprehensive metabolomic analysis. For the first time in the present study, we juxtapose the most commonly employed mass spectrometry-based analysis methodologies and directly compare the resultant coverages of detected compounds in exhaled breath condensate in order to guide methodology choices for exhaled breath condensate analysis studies. Four methods were explored to broaden the range of measured compounds across both the volatile and non-volatile domain. Liquid phase sampling with polyacrylate Solid-Phase MicroExtraction fiber, liquid phase extraction with a polydimethylsiloxane patch, and headspace sampling using Carboxen/Polydimethylsiloxane Solid-Phase MicroExtraction (SPME) followed by gas chromatography mass spectrometry were tested for the analysis of volatile fraction. Hydrophilic interaction liquid chromatography and reversed-phase chromatography high performance liquid chromatography mass spectrometry were used for analysis of non-volatile fraction. We found that liquid phase breath condensate extraction was notably superior compared to headspace extraction and differences in employed sorbents manifested altered metabolite coverages. The most pronounced effect was substantially enhanced metabolite capture for larger, higher-boiling compounds using polyacrylate SPME liquid phase sampling. The analysis of the non-volatile fraction of breath condensate by hydrophilic and reverse phase high performance liquid chromatography mass spectrometry indicated orthogonal metabolite coverage by these chromatography modes. We found that the metabolite coverage could be enhanced significantly with the use of organic solvent as a device rinse after breath sampling to collect the non-aqueous fraction as opposed to neat breath condensate sample. Here, we show the detected ranges of compounds in each case and provide a practical guide for methodology selection for optimal detection of specific compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
24241-U Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
25461 Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
24246 Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
25467 Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
54041-U Carboxen® 1000 Reversible SPE Tube, 200 mg/mL, pk 30
29753-U Carboxen® 569, O.D. × L 1/4 in. × 3 1/2, glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
29534-U Carboxen® 569, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
54293-U Carboxen® 572 for Tobacco Smoke, Adsorbent Bed Wt: (300 mg), cartridge volume 3 mL, pkg of 10 ea
54294-U Carboxen® 572 for Tobacco Smoke, Adsorbent Bed Wt: 300 mg, cartridge size 3 mL, pkg of 50 ea
10263 Carboxen® Adsorbent, matrix Carboxen® 563, 20-45 mesh, bottle of 10 g
10264 Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, bottle of 10 g
11324-U Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
10269 Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
11048-U Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
11072-U Carboxen® Adsorbent, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
10477-U Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
10478-U Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
10471 Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
SU860099
Headspace vial, screw top, rounded bottom (vial only), volume 10 mL, clear glass vial, thread 18, O.D. × H 22.5 mm × 46 mm, pkg of 100 ea
SU860101
Magnetic Screw Cap for Headspace Vials, 18 mm thread, PTFE/silicone septum (white PTFE/tranparent blue silicone), septum thickness 1.3 mm, pkg of 100 ea
23446-U ORBO-93 Sorbent Tube, matrix Carboxen® 1000, ORBO tube I.D. × L 6 mm × 95 mm, Bed B 90 mg, Bed A 180 mg, 60/80 mesh, pkg of 25 ea
20355 ORBO 78 HBR on Carboxen® 564 specially cleaned (20/45), 400/200 mg, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
20358 ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
28718-U Tenax® TA / Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4 in. × 3 1/2 in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea