EMAIL THIS PAGE TO A FRIEND
The Canadian veterinary journal = La revue veterinaire canadienne
Cardiovascular effects of constant rate infusions of lidocaine, lidocaine and dexmedetomidine, and dexmedetomidine in dogs anesthetized at equipotent doses of sevoflurane.
PMID 28698692
Abstract
This study evaluated the cardiovascular effects of a constant rate infusion (CRI) of lidocaine, lidocaine and dexmedetomidine, and dexmedetomidine in dogs anesthetized with sevoflurane at equipotent doses. Treatments consisted of T1-Lidocaine [loading dose 2 mg/kg body weight (BW), IV, and CRI of 100 μg/kg BW per min] at 1.4% end-tidal of sevoflurane (FE