EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Regulatory, integrative and comparative physiology
Functional and neurochemical characterization of angiotensin type 1A receptor-expressing neurons in the nucleus of the solitary tract of the mouse.
PMID 28701322
Abstract
Angiotensin II acts via two main receptors within the central nervous system, with the type 1A receptor (AT