EMAIL THIS PAGE TO A FRIEND
Journal of neurophysiology
Differential contributions to the interception of occluded ballistic trajectories by the temporoparietal junction, area hMT/V5+, and the intraparietal cortex.
PMID 28701531
Abstract
The ability to catch objects when transiently occluded from view suggests their motion can be extrapolated. Intraparietal cortex (IPS) plays a major role in this process along with other brain structures, depending on the task. For example, interception of objects under Earth's gravity effects may depend on time-to-contact predictions derived from integration of visual signals processed by hMT/V5+ with a priori knowledge of gravity residing in the temporoparietal junction (TPJ). To investigate this issue further, we disrupted TPJ, hMT/V5+, and IPS activities with transcranial magnetic stimulation (TMS) while subjects intercepted computer-simulated projectile trajectories perturbed randomly with either hypo- or hypergravity effects. In