EMAIL THIS PAGE TO A FRIEND
Scientific reports
Recycling and Self-Healing of Polybenzoxazines with Dynamic Sulfide Linkages.
PMID 28701718
Abstract
In this work, a recycling and self-healing strategy for polybenzoxazines through both S-S bond cleavage-reformation reaction and supramolecular attractions is described. Both recyclable and self-healable polybenzoxazines can be prepared from low cost chemicals with a simple procedure in only 30 minutes. For this purpose, inverse vulcanization of poly(propylene oxide)benzoxazine (PPOB) and diallybenzoxazine (B-al) with elemental sulfur was performed at 185 °C. The obtained cross-linked polymer films exhibited thermally driven recycling ability up to 5 cycles. Moreover, the self-healing ability of a test specimen was shown. Spectral characterizations, thermal stability and fracture toughness of the films were investigated after each recycling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
133027
Bisphenol A, 97%
C15H16O2