EMAIL THIS PAGE TO A FRIEND
Journal of ginseng research
Quality and characteristics of fermented ginseng seed oil based on bacterial strain and extraction method.
PMID 28701887
Abstract
In this study, the fermentation of ginseng seeds was hypothesized to produce useful physiologically-active substances, similar to that observed for fermented ginseng root. Ginseng seed was fermented using The ginseng seed was fermented inoculating 1% of each strain on sterilized ginseng seeds and incubating the seeds at 30°C for 24 h. Oil was extracted from the fermented ginseng seeds using compression extraction, solvent extraction, and supercritical fluid extraction. The color of the fermented ginseng seed oil did not differ greatly according to the fermentation or extraction method. The highest phenolic compound content recovered with the use of supercritical fluid extraction combined with fermentation using the
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5157
Campesterol, ~65%
C28H48O