EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Arsanilic acid modified superparamagnetic iron oxide nanoparticles for Purification of alkaline phosphatase from hen's egg yolk.
PMID 28704722
Abstract
Recent studies of magnetic carrier technology have focused on its applications in separation and purification technologies, due to easy separation of the target from the reaction medium by applying an external magnetic field. In the present study, Fe
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
440183
(3-Chloropropyl)trimethoxysilane, ≥97%
C6H15ClO3Si
239690
Succinic anhydride, ≥99% (GC)
C4H4O3