EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Gastrointestinal and liver physiology
Augmentation of cGMP/PKG pathway and colonic motility by hydrogen sulfide.
PMID 28705807
Abstract
Hydrogen sulfide (H