EMAIL THIS PAGE TO A FRIEND
Nature communications
Structural and functional characterization of a DARPin which inhibits Ras nucleotide exchange.
PMID 28706291
Abstract
Ras mutations are the oncogenic drivers of many human cancers and yet there are still no approved Ras-targeted cancer therapies. Inhibition of Ras nucleotide exchange is a promising new approach but better understanding of this mechanism of action is needed. Here we describe an antibody mimetic, DARPin K27, which inhibits nucleotide exchange of Ras. K27 binds preferentially to the inactive Ras GDP form with a K