EMAIL THIS PAGE TO A FRIEND
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology
Relaxant and anti-inflammatory effect of two thalidomide analogs as PDE-4 inhibitors in pregnant rat uterus.
PMID 28706457
Abstract
The aim of this study was to evaluate the relaxant and anti-inflammatory effects of two thalidomide analogs as phosphodiesterase-4 (PDE-4) inhibitors in pregnant rat uterus. Uteri from Wistar female rats were isolated at 19 day of pregnancy. Uterine samples were used in functional studies to evaluate the inhibitory effects of the thalidomide analogs, methyl 3-(4-nitrophthalimido)-3-(3,4-dimethoxyphenyl)-propanoate (4NO2PDPMe) and methyl 3-(4-aminophthalimido)-3-(3,4-dimethoxyphenyl)-propanoate (4APDPMe), on prostaglandin-F2α (PGF2α)-induced phasic, K