Environmental research
Bisphenol A induces COX-2 through the mitogen-activated protein kinase pathway and is associated with levels of inflammation-related markers in elderly populations.
PMID 28709031
Bisphenol A (BPA) is a well-known endocrine-disrupting chemical, and it is one of the highest volume chemicals produced worldwide. Even though several in vivo and in vitro studies showed positive associations of BPA exposure with pro-inflammatory cytokines such as tumor necrosis factor-α and interleukin (IL)-6, the mechanism by which BPA induces inflammation is unclear. We investigated the mechanism by which BPA induces inflammation (expression of inflammation-related genes, changes in oxidative stress, and cell proliferation and migration) and evaluated the effect of BPA exposure on inflammation-related markers in epidemiologic studies using repeat urine and serum samples from elderly subjects. BPA induced COX-2 expression via nuclear translocation of NF-κB and activation of mitogen-activated protein kinase (MAPK) by phosphorylation of ERK1/2 and enhanced the migration of lung cancer A549 and breast cancer MDAMB-231 cells. In two epidemiologic studies, we detected associations of BPA with six inflammation-related markers (WBC, CRP, IL-10, ALT, AST, and γ-GTP levels). Our findings probably suggest that BPA exposure induces inflammation and exacerbates tumorigenesis.