EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Application of magnetic OMS-2 in sequencing batch reactor for treating dye wastewater as a modulator of microbial community.
PMID 28711831
Abstract
The potential and mechanism of synthesized magnetic octahedral molecular sieve (Fe
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
330825
Manganese(II) acetate, 98%
C4H6MnO4