EMAIL THIS PAGE TO A FRIEND
Frontiers in plant science
Isolation of an
PMID 28713410
Abstract
Abscisic acid (ABA) transporters are essential for the transport of ABA from its sites of synthesis to its multiple sites of action within plants and are key players in plant stress responses. Despite their importance, there is limited information on ABA transporters in crop plants. In this study, we isolated and characterized an