EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Selection of an optimal promoter for gene transfer in normal B cells.
PMID 28713922
Abstract
Gene transfer into normal quiescent human B cells is a challenging procedure. The present study aimed to investigate whether it is possible to increase the levels of transgene expression by using various types of promoters to drive the expression of selected genes‑of‑interest. To produce lentiviral particles, the present study used the 2nd generation psPAX2 packaging vector and the vesicular stomatitis virus ‑expressing envelope vector pMD2.G. Subsequently, lentiviral vectors were generated containing various promoters, including cytomegalovirus (CMV), elongation factor‑1 alpha (EF1α) and spleen focus‑forming virus (SFFV). The present study was unable to induce satisfactory transduction efficiency in quiescent normal B cells; however, infection of normal B cells with Epstein‑Barr virus resulted in increased susceptibility to lentiviral transduction. In addition, the SFFV promoter resulted in a higher level of transgene expression compared with CMV or EF1α promoters. As a proof‑of concept that this approach allows for stable gene expression in normal B cells, the present study used bicistronic lentiviral vectors with genes encoding fluorescent reporter proteins, as well as X‑box binding protein‑1 and binding immunoglobulin protein.