EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Functionalized superparamagnetic nanoparticles with a polymer containing β-cyclodextrin for the extraction of sertraline hydrochloride in biological samples.
PMID 28714248
Abstract
In this study, a novel magnetic nanoadsorbent was synthesized by grafting β-cyclodextrin onto the modified surface of Fe