The Journal of general virology
Hepatitis B virus X protein activates E3 ubiquitin ligase Siah-1 to control virus propagation via a negative feedback loop.
PMID 28714848
The seven in absentia homologue 1 (Siah-1) protein is an E3 ubiquitin ligase that induces ubiquitin-dependent proteasomal degradation of HBx, the principal regulatory protein of hepatitis B virus (HBV); however, its role in HBV propagation remains unknown. Here, we found that HBx upregulates Siah-1 levels in HepG2 but not in Hep3B cells, in which p53 is absent. For this effect, HBx sequentially activated ataxia telangiectasia mutated kinase and checkpoint kinase 2 via phosphorylation at the Ser-1981 and Thr-68 residues, respectively, which led to the activation of p53 via phosphorylation at the Ser-15 and Ser-20 residues. As a result, HBx was heavily ubiquitinated by Siah-1 and degraded by the ubiquitin-proteasome system in HepG2 cells, whereas this effect was marginal or undetectable in Hep3B cells. Knock-down of p53 in HepG2 cells downregulated Siah-1 levels and subsequently upregulated HBx levels, whereas ectopic p53 expression in Hep3B cells upregulated Siah-1 levels and subsequently downregulated HBx levels. In addition, Siah-1 knock-down impaired the ubiquitination and proteasomal degradation of HBx in HepG2 cells, whereas ectopic Siah-1 expression induced ubiquitin-dependent proteasomal degradation of HBx in Hep3B cells. The effects of HBx on p53 and Siah-1 were exactly reproduced in a 1.2-mer HBV replicon system, mimicking the natural course of HBV infection. In particular, Siah-1 knock-down upregulated the levels of HBx derived from the HBV replicon, resulting in an increase in HBV production. In conclusion, HBx modulates its own protein level via a negative feedback loop involving p53 and Siah-1 to control HBV propagation.
Related Materials
Product #
Molecular Formula
Add to Cart
EHU071391 MISSION® esiRNA, esiRNA human SIAH1 (esiRNA1)