EMAIL THIS PAGE TO A FRIEND
Nature neuroscience
Gut microbiota is critical for the induction of chemotherapy-induced pain.
PMID 28714953
Abstract
Chemotherapy-induced pain is a dose-limiting condition that affects 30% of patients undergoing chemotherapy. We found that gut microbiota promotes the development of chemotherapy-induced mechanical hyperalgesia. Oxaliplatin-induced mechanical hyperalgesia was reduced in germ-free mice and in mice pretreated with antibiotics. Restoring the microbiota of germ-free mice abrogated this protection. These effects appear to be mediated, in part, by TLR4 expressed on hematopoietic cells, including macrophages.
Related Materials