EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Self-assembly of zinc hydroxystannate on amorphous hydrous TiO
PMID 28715749
Abstract
Zinc hydroxystannate (ZHS) was fabricated on the surface of amorphous hydrous TiO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14505
Sodium stannate, 42-45% SnO2 basis
Na2O3Sn