EMAIL THIS PAGE TO A FRIEND
Nanoscale research letters
Substitutional Doping for Aluminosilicate Mineral and Superior Water Splitting Performance.
PMID 28715878
Abstract
Substitutional doping is a strategy in which atomic impurities are optionally added to a host material to promote its properties, while the geometric and electronic structure evolution of natural nanoclay mineral upon substitutional metal doping is still ambiguous. This paper first designed an efficient lanthanum (La) doping strategy for nanotubular clay (halloysite nanotube, HNT) through the dynamic equilibrium of a substitutional atom in the presence of saturated AlCl