EMAIL THIS PAGE TO A FRIEND
The EMBO journal
Type I interferon is required for T helper (Th) 2 induction by dendritic cells.
PMID 28716804
Abstract
Type 2 inflammation is a defining feature of infection with parasitic worms (helminths), as well as being responsible for widespread suffering in allergies. However, the precise mechanisms involved in T helper (Th) 2 polarization by dendritic cells (DCs) are currently unclear. We have identified a previously unrecognized role for type I IFN (IFN-I) in enabling this process. An IFN-I signature was evident in DCs responding to the helminth