EMAIL THIS PAGE TO A FRIEND
Journal of lipid research
DOCK2 deficiency mitigates HFD-induced obesity by reducing adipose tissue inflammation and increasing energy expenditure.
PMID 28716822
Abstract
Obesity is the major risk factor for type 2 diabetes, cardiovascular disorders, and many other diseases. Adipose tissue inflammation is frequently associated with obesity and contributes to the morbidity and mortality. Dedicator of cytokinesis 2 (DOCK2) is involved in several inflammatory diseases, but its role in obesity remains unknown. To explore the function of DOCK2 in obesity and insulin resistance, WT and DOCK2-deficient (DOCK2