EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Creation and Preliminary Characterization of Pregnane X Receptor and Constitutive Androstane Receptor Knockout Rats.
PMID 28716828
Abstract
The nuclear receptors pregnane X receptor (PXR) and constitutive androstane receptor (CAR) are closely related transcription factors that regulate the expression of phase I (cytochrome P450s) and phase II metabolizing enzymes and transporter genes in response to stimulation from xenobiotics, including prescription drugs. PXR and CAR knockout and humanized mouse models have proven useful. However, the rat being bigger in size is a preferred model system for studying drug metabolism and pharmacokinetics. Here, we report the creation and preliminary characterization of PXR and CAR knockout rats and PXR/CAR double knockout rats. Whereas the expression of phase I and II enzymes and transporter genes were not upregulated by nuclear receptor-specific agonists pregnenlone-16
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T1443
TCPOBOP, ≥98% (HPLC), solid
C16H8N2O2Cl4