EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Prolonged force depression after mechanically demanding contractions is largely independent of Ca
PMID 28716970
Abstract
Increased production of reactive oxygen/nitrogen species (ROS) and impaired cellular Ca