EMAIL THIS PAGE TO A FRIEND
Frontiers in plant science
Ethanol Enhances High-Salinity Stress Tolerance by Detoxifying Reactive Oxygen Species in
PMID 28717360
Abstract
High-salinity stress considerably affects plant growth and crop yield. Thus, developing techniques to enhance high-salinity stress tolerance in plants is important. In this study, we revealed that ethanol enhances high-salinity stress tolerance in