EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Environmentally benign dry-gel conversions of Zr-based UiO metal-organic frameworks with high yield and the possibility of solvent re-use.
PMID 28719671
Abstract
Herein we report an alternative synthesis method for Zr-based UiO metal-organic frameworks (MOFs), namely dry-gel conversion (DGC). It was possible to synthesize nano- to microsized UiO-66, UiO-66-NH
Related Materials