EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
High photocatalytic activity of hierarchical SiO
PMID 28719847
Abstract
Ongoing research activities are targeted to explore high photocatalytic activity of TiO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
408107
[2-(Methacryloyloxy)ethyl]trimethylammonium chloride solution, 75 wt. % in H2O
C9H18ClNO2