EMAIL THIS PAGE TO A FRIEND
Malaria journal
Effect of seasonal malaria chemoprevention on the acquisition of antibodies to Plasmodium falciparum antigens in Ouelessebougou, Mali.
PMID 28720100
Abstract
Seasonal malaria chemoprevention (SMC) is a new strategy to reduce malaria burden in young children in Sahelian countries. It consists of the administration of full treatment courses of sulfadoxine-pyrimethamine plus amodiaquine to children at monthly intervals during the malaria season. However, it is not clear if there is a cumulative effect of SMC over time on acquisition of antibodies to malaria antigens. A cross-sectional serosurvey was carried out 1 month after the last dose of SMC in 2016. Children aged 3-4 years were randomly selected from areas where SMC was given for 1, 2 or 3 years during the malaria season. Children in the areas where SMC had been implemented for 1 year but who failed to receive SMC were used as comparison group. Antibody extracted from dry blood spots was used to measure IgG levels to CSP, MSP-1 The prevalence of antibodies to AMA-1 were high and similar in children who received SMC for 1, 2 or 3 years and also when compared to those who never received SMC (96.3 vs 97.5%, adjusted OR = 0.99, 95% CI 0.33-2.97, p = 0.99). The prevalence of antibodies to MSP-1 SMC reduced seropositivity to MSP-1
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9187 SIGMAFAST OPD, tablet