EMAIL THIS PAGE TO A FRIEND
Journal of cellular biochemistry
Breast cancer stem cells are involved in Trastuzumab resistance through the HER2 modulation in 3D culture.
PMID 28722778
Abstract
Breast cancer human cells culture as spheroids develop autophagy and apoptosis, which promotes Trastuzumab resistance in HER2 overexpressing cells. Our aim was to study the association of the hostile environment developed in 3D with the breast cancer stem cells population and the HER2 modulation. Human mammary adenocarcinoma cell lines were cultured as spheroids using the hanging drop method. We generated hypoxia conditions by using a hypoxic chamber and CoCl