EMAIL THIS PAGE TO A FRIEND
ACS sensors
Augmented Reality for Real-Time Detection and Interpretation of Colorimetric Signals Generated by Paper-Based Biosensors.
PMID 28723111
Abstract
Colorimetric tests are becoming increasingly popular in point-of-need analyses due to the possibility of detecting the signal with the naked eye, which eliminates the utilization of bulky and costly instruments only available in laboratories. However, colorimetric tests may be interpreted incorrectly by nonspecialists due to disparities in color perception or a lack of training. Here we solve this issue with a method that not only detects colorimetric signals but also interprets them so that the test outcome is understandable for anyone. It consists of an augmented reality (AR) app that uses a camera to detect the colored signals generated by a nanoparticle-based immunoassay, and that yields a warning symbol or message when the concentration of analyte is higher than a certain threshold. The proposed method detected the model analyte mouse IgG with a limit of detection of 0.3 μg mL