EMAIL THIS PAGE TO A FRIEND
Assay and drug development technologies
Development of Three Orthogonal Assays Suitable for the Identification and Qualification of PIKfyve Inhibitors.
PMID 28723271
Abstract
FYVE-type zinc finger-containing phosphoinositide kinase (PIKfyve) catalyzes the formation of phosphatidylinositol 3,5-bisphosphate (PI(3,5)P